Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATDC Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991982
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1982 100 μg
50-199-1983 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1982 Supplier Rockland Immunochemicals Supplier No. 600401B15
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with over-expressed, acetylated and non-acetylated (at K116) ATDC protein. A BLAST analysis was used to suggest cross-reactivity with ATDC from human, horse, bovine, chimpanzee and macaque based on a 100% homology with the immunizing sequence. Partial reactivity is expected against rat and mouse ATDC based on 92% homology with the immunizing sequence. Cross-reactivity with ATDC from other sources has not been determined.

The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype.

Specifications

Antigen ATDC
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Trim29
Gene Accession No. Q14134
Gene Alias 1110047J21Rik; 2810431N19Rik; 4732461M22Rik; AI119726; Ataxia telangiectasia group D-associated protein; ataxia-telangiectasia group D-associated protein; ATDC; FLJ36085; TRIM29; tripartite motif containing 29; tripartite motif protein 29; tripartite motif protein TRIM29; tripartite motif-containing 29; Tripartite motif-containing protein 29
Gene Symbols Trim29
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100071461, 23650, 451615, 525062, 705796
Target Species Bovine, Chimpanzee, Equine, Human, Rhesus Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.