Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atenolol, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18600809 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle

A1648, 29122-68-7, C14H22N2O3

Spectrum™ Chemical Atenolol, USP A receptor antagonist that belongs to the beta blocker group. It is a drug used to treat cardiovascular diseases by slowing the heart beat down and in turn reducing the strain on the heart. It is used for several conditions that include supraventricular tachycardia, hypertension, angina and acute myocardial infarction. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 29122-68-7
Molecular Formula C14H22N2O3
Molecular Weight (g/mol) 266.34
InChI Key METKIMKYRPQLGS-UHFFFAOYNA-N
IUPAC Name 2-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)acetamide
SMILES CC(C)NCC(O)COC1=CC=C(CC(N)=O)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Melting Range 152-156.5°C
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.