Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATF1 Monoclonal Antibody (7F8)
GREENER_CHOICE

Catalog No. PIMA549203
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549203 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549203 Supplier Invitrogen™ Supplier No. MA549203
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human ATF1 shares 91% amino acid (aa) sequence identity with mouse ATF1. Positive Control - WB: human HepG2 whole cell, human Hela whole cell, human SW620 whole cell. IHC: human lung cancer tissue, human rectal cancer tissue, human rectal cancer tissue. ICC/IF: A431 cell. Flow: K562 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes an activating transcription factor, which belongs to the ATF subfamily and bZIP (basic-region leucine zipper) family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. This protein is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and cyclin-dependent kinase 3 (cdk-3). Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation. Fusion of this gene and FUS on chromosome 16 or EWSR1 on chromosome 22 induced by translocation generates chimeric proteins in angiomatoid fibrous histiocytoma and clear cell sarcoma. This gene has a pseudogene on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7F8
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene ATF1
Gene Accession No. P18846
Gene Alias AATase 1; Activating transcription factor 1; alcohol O-acetyltransferase; Alcohol O-acetyltransferase 1; ATF1; Atf-1; cAMP-dependent transcription factor ATF-1; cyclic AMP-dependent transcription factor ATF-1; EWS-ATF1; FUS/ATF-1; Protein TREB36; TCR-ATF1; TREB36; Unknown (protein for MGC:64099); wu:fc92c07; YOR377W; zgc:64099
Gene Symbols ATF1
Host Species Mouse
Immunogen E.coli-derived human ATF1 recombinant protein (Position: M1-V271).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 466
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.