Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541490 Supplier Invitrogen™ Supplier No. PA541490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KNREAARECR RKKKEYVKCL ENRVAVLENQ NKTLIEELKT LKDLYSNKSV Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Sheep: 77%; Zebrafish: 100%.

This gene encodes an activating transcription factor, which belongs to the ATF subfamily and bZIP (basic-region leucine zipper) family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. This protein is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and cyclin-dependent kinase 3 (cdk-3). Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation. Fusion of this gene and FUS on chromosome 16 or EWSR1 on chromosome 22 induced by translocation generates chimeric proteins in angiomatoid fibrous histiocytoma and clear cell sarcoma. This gene has a pseudogene on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATF1
Gene Accession No. P18846
Gene Alias AATase 1; Activating transcription factor 1; alcohol O-acetyltransferase; Alcohol O-acetyltransferase 1; ATF1; Atf-1; cAMP-dependent transcription factor ATF-1; cyclic AMP-dependent transcription factor ATF-1; EWS-ATF1; FUS/ATF-1; Protein TREB36; TCR-ATF1; TREB36; Unknown (protein for MGC:64099); wu:fc92c07; YOR377W; zgc:64099
Gene Symbols ATF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ATF1 (aa 222-271).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 466
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.