Learn More
ATF1 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | ATF1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | ATF1 |
| Gene Accession No. | P18846 |
| Gene Alias | AATase 1; Activating transcription factor 1; alcohol O-acetyltransferase; Alcohol O-acetyltransferase 1; Atf1; Atf-1; BOS_5060; cAMP-dependent transcription factor ATF-1; Cyclic AMP-dependent transcription factor ATF-1; EWS-ATF1; FUS/ATF-1; LOC100340999; Protein TREB36; TCR-ATF1; TREB36; Unknown (protein for MGC:64099); wu:fc92c07; YOR377W; zgc:64099 |
| Gene Symbols | ATF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.