Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0517R

Description
This gene encodes an activating transcription factor, which belongs to the ATF subfamily and bZIP (basic-region leucine zipper) family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. This protein is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and cyclin-dependent kinase 3 (cdk-3). Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation. Fusion of this gene and FUS on chromosome 16 or EWSR1 on chromosome 22 induced by translocation generates chimeric proteins in angiomatoid fibrous histiocytoma and clear cell sarcoma. This gene has a pseudogene on chromosome 6.Specifications
ATF1 | |
Polyclonal | |
Unconjugated | |
ATF1 | |
AATase 1; Activating transcription factor 1; alcohol O-acetyltransferase; Alcohol O-acetyltransferase 1; Atf1; Atf-1; BOS_5060; cAMP-dependent transcription factor ATF-1; Cyclic AMP-dependent transcription factor ATF-1; EWS-ATF1; FUS/ATF-1; LOC100340999; Protein TREB36; TCR-ATF1; TREB36; Unknown (protein for MGC:64099); wu:fc92c07; YOR377W; zgc:64099 | |
Rabbit | |
Protein A | |
RUO | |
466 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P18846 | |
ATF1 | |
KLH conjugated synthetic peptide derived from human ATF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction