Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    ATF2 Antibody (PCRP-ATF2-1B4), DyLight 488, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314001G
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
ATF2 Monoclonal antibody specifically detects ATF2 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| ATF2 | |
| Monoclonal | |
| DyLight 488 | |
| activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 | |
| Recombinant full-length human ATF2 protein (Uniprot: P15336) | |
| 0.1 mL | |
| Cancer, Chromatin Research, Immunology, Innate Immunity, Transcription Factors and Regulators | |
| 1386 | |
| Store at 4°C in the dark. | |
| IgG2b | 
| Western Blot, Flow Cytometry, Immunofluorescence | |
| PCRP-ATF2-1B4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction