Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF2, p Thr71 Antibody (103C411.2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056594FR
Description
ATF2 Monoclonal specifically detects ATF2 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
ATF2 (p Thr71) | |
Monoclonal | |
DyLight 680 | |
P15336 | |
ATF2 | |
This antibody was generated by immunizing mice with a synthetic peptide containing phosphorylated threonine at position 71 of human ATF-2. | |
0.1 mL | |
Cancer, Chromatin Research, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
1386 | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
103C411.2 | |
Western Blot, Flow Cytometry | |
activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This monoclonal antibody is specific for phospho (Thr71) ATF2. It cannot be used to test for non-phosphorylated ATF2. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction