Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF2, p Thr71 Antibody (103C411.2), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen ATF2 (p Thr71)
Clone 103C411.2
Concentration 0.5 mg/mL
Dilution Western Blot 2 ug/ml, Flow Cytometry 1:10-1:1000
Applications Western Blot, Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056594T
SDP
View Documents
Novus Biologicals
NB10056594SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056594
SDP
View Documents
Novus Biologicals
NB10056594
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

ATF2 Monoclonal specifically detects ATF2 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

ATF2 (p Thr71)
0.5 mg/mL
Western Blot, Flow Cytometry
Unconjugated
Mouse
Cancer, Chromatin Research, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
P15336
1386
This antibody was generated by immunizing mice with a synthetic peptide containing phosphorylated threonine at position 71 of human ATF-2.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
103C411.2
Western Blot 2 ug/ml, Flow Cytometry 1:10-1:1000
Monoclonal
Purified
RUO
Human
activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558
ATF2
IgG1
Protein G purified
This monoclonal antibody is specific for phospho (Thr71) ATF2. It cannot be used to test for non-phosphorylated ATF2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.