Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF2, p Thr71 Antibody (4F2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | ATF2 |
---|---|
Clone | 4F2 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
ATF2 Monoclonal antibody specifically detects ATF2 in Human samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
ATF2 | |
Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 | |
A synthesized peptide derived from Human ATF2 [p Thr71] [UniProt P15336] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4F2 | |
Western Blot, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1386 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title