Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF3 Antibody (CL1685), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP23448925UL
Description
ATF3 Monoclonal specifically detects ATF3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ATF3 | |
Monoclonal | |
Unconjugated | |
P18847 | |
ATF3 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MMLQHPGQVSASEVSASAIVPCLSPPGSLVFEDFANLTPFVKEELRFAIQNKHLCHRMSSALESVTVSDRPLGVSITKAEVAPEEDERKKRRRERNKIAAAKCRNKKKEKTEC | |
25 μL | |
Cancer, Transcription Factors and Regulators | |
467 | |
Human | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CL1685 | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 2-10 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction