Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF3 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP102935UV
Description
ATF3 Polyclonal specifically detects ATF3 in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATF3 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3 | |
Rabbit | |
Affinity Purified | |
Cancer, Transcription Factors and Regulators | |
467.0 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ATF3 | |
A synthetic peptide corresponding to a region of human ATF3 (within residues 100-150). [UniProtNo. P18847] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Mammalia | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction