Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF4 Antibody (S360A-24), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242176F

Catalog No. NBP242176F


Only null left
Add to Cart

Description

Description

ATF4 Monoclonal specifically detects ATF4 in Human, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ATF4
Monoclonal
FITC
PBS with 0.05% Sodium Azide
Activating transcription factor 4, activating transcription factor 4 (tax-responsive enhancer element B67), cAMP-dependent transcription factor ATF-4, cAMP-responsive element-binding protein 2, CREB-2DNA-binding protein TAXREB67, cyclic AMP-dependent transcription factor ATF-4, Cyclic AMP-responsive element-binding protein 2, TaxREB67, TAXREB67CREB2cAMP response element-binding protein 2, Tax-responsive enhancer element-binding protein 67, TXREB
Mouse
Protein G purified
Lipid and Metabolism, Phospho Specific, Unfolded Protein Response, Unfolded Protein Response (UPR)
468
Human, Rat
Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S360A-24
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
P18848, NP_001666.2
ATF4
Fusion protein amino acids 25-327 of human ATF4. 86% identical to rat, and 85% identical to mouse. <50% identity with ATF5.
0.1 mL
Primary
Less then 50% identity with ATF5.Detects approx 60kDa. Does not cross-react with ATF5.
Store at 4C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.