Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF4 Antibody (S360A-24), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242176H
Description
ATF4 Monoclonal specifically detects ATF4 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ATF4 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Activating transcription factor 4, activating transcription factor 4 (tax-responsive enhancer element B67), cAMP-dependent transcription factor ATF-4, cAMP-responsive element-binding protein 2, CREB-2DNA-binding protein TAXREB67, cyclic AMP-dependent transcription factor ATF-4, Cyclic AMP-responsive element-binding protein 2, TaxREB67, TAXREB67CREB2cAMP response element-binding protein 2, Tax-responsive enhancer element-binding protein 67, TXREB | |
Mouse | |
Protein G purified | |
Lipid and Metabolism, Phospho Specific, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
468 | |
Human, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
S360A-24 | |
Western Blot, Immunohistochemistry | |
P18848, NP_001666.2 | |
ATF4 | |
Fusion protein amino acids 25-327 of human ATF4. 86% identical to rat, and 85% identical to mouse. <50% identity with ATF5. | |
0.1 mL | |
Primary | |
Less then 50% identity with ATF5.Detects approx 60kDa. Does not cross-react with ATF5. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction