Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF4, p Ser245 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10081802
Description
ATF4 Polyclonal specifically detects ATF4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATF4 (p Ser245) | |
Polyclonal | |
Unconjugated | |
PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
Activating transcription factor 4, activating transcription factor 4 (tax-responsive enhancer element B67), cAMP-dependent transcription factor ATF-4, cAMP-responsive element-binding protein 2, CREB-2DNA-binding protein TAXREB67, cyclic AMP-dependent transcription factor ATF-4, Cyclic AMP-responsive element-binding protein 2, TaxREB67, TAXREB67CREB2cAMP response element-binding protein 2, Tax-responsive enhancer element-binding protein 67, TXREB | |
Rabbit | |
45 kDa | |
0.05 mL | |
Phospho Specific | |
468 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
P18848 | |
ATF4 | |
The antiserum was produced against synthesized phosphopeptide derived from human ATF4 around the phosphorylation site of serine 245 (N-R-SP-L-P). | |
Affinity Purified | |
RUO | |
Primary | |
Detects endogenous levels of ATF4 only when phosphorylated at serine 245. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction