Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF4 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993592
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3592 100 μL
50-199-3593 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3592 Supplier Active Motif Supplier No. 61655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.

Specifications

Antigen ATF4
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.5
Gene Atf4
Gene Accession No. P18848
Gene Alias Activating transcription factor 4; activating transcription factor 4 (tax-responsive enhancer element B67); activating transcription factor 4a; activating transcription factor 4b1 (tax-responsive enhancer element B67); activating transcription factor ATF-4; ATF 4; atf4; Atf-4; atf4 {ECO:0000312; ATF4 protein; atf4a; atf4b1; BOS_6087; C/ATF; C/EBP-related ATF; cAMP response element-binding protein 2; cAMP-dependent transcription factor ATF-4; cAMP-responsive element-binding protein 2; CREB 2; CREB2; CREB-2; Cyclic AMP-dependent transcription factor ATF-4; Cyclic AMP-responsive element-binding protein 2; DNA-binding protein TAXREB67; OTTHUMP00000199130; rATF-4; RGD:621863}; tax-re; TaxREB67; TaxREB67 homolog; tax-responsive enhancer element B67; Tax-responsive enhancer element-binding protein 67; Tax-responsive enhancer element-binding protein 67 homolog; TXREB; wu:fj01c08; wu:fk30b04; zgc:85951
Gene Symbols Atf4
Host Species Rabbit
Immunogen This antibody was raised against a recombinant protein to human ATF4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 468
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.