Learn More
ATF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 108351AP150UL

Description
This antibody is a rabbit polyclonal antibody raised against full length human ATF4 antigen. The antibody recognizes the 38 kDa ATF4 protein and its phosphorylated forms (50 kDa). ATF4 can bind DNA as a homodimer and as a heterodimer. ATF4 is ubiquitinate
This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.Specifications
ATF4 | |
Polyclonal | |
Unconjugated | |
ATF4 | |
ATF4, CREB 2, CREB2, DNA binding protein TAXREB67, TAXREB67, TXREB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11911, 468, 79255 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18848, Q06507, Q9ES19 | |
ATF4 | |
ATF4 Fusion Protein Ag1279 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.