Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF5 Rabbit anti-Monkey, Rat, Human, Polyclonal, MilliporeSigma™

Catalog No. ABE2614MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2614MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2614MI Supplier MilliporeSigma Supplier No. ABE2614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ATF5 is a highly specific rabbit polyclonal antibody that targets Cyclic AMP-dependent transcription factor ATF-5 and has been tested in Immunocytochemistry, Immunofluorescence, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF5
Applications Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ATFS,ATFX
Host Species Rabbit
Immunogen A linear peptide corresponding to 18 amino acids from the N-terminal half of human Cyclic AMP-dependent transcription factor ATF-5.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_036200
Test Specificity This polyclonal antibody targets a sequence in the N-terminal region and detects ATF5 by immunocytochemstry.
Target Species Monkey, Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.