Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF5 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310652100UL
Description
ATF5 Polyclonal specifically detects ATF5 in Rat samples. It is validated for Western Blot.Specifications
ATF5 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
activating transcription factor 5FLJ34666, ATFXHMFN0395, cAMP-dependent transcription factor ATF-5, cyclic AMP-dependent transcription factor ATF-5, Transcription factor ATFx | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Rat ATF5 (NP_758839). Peptide sequence EGEALEGECQGLEARNRELRERAESVEREIQYVKDLLIEVYKARSQRTRS | |
100 μg | |
Cell Cycle and Replication | |
22809 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction