Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF6 Antibody (70B1413.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1402560.1MG
Description
ATF6 Monoclonal specifically detects ATF6 in Human, Mouse, Rat, Porcine, Plant, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), CyTOF-ready, Knockout Validated.Specifications
ATF6 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 3 - 5 ug/ml, Chromatin Immunoprecipitation 1:10 - 1:500, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, CyTOF-ready | |
P18850 | |
ATF6 | |
This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6. | |
0.1 mg | |
Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
22926 | |
Human, Mouse, Rat, Pig, Fish, Hamster, Rabbit | |
Purified |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
70B1413.1 | |
Unconjugated | |
TBS with 0.05% Sodium Azide | |
activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This ATF6 antibody detects both the full length and the cleaved/active protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction