Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF6 Antibody (70B1413.1), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP140256MFV610
Description
ATF6 Monoclonal antibody specifically detects ATF6 in Human, Mouse, Rat, Porcine, Plant, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
ATF6 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Plant, Rabbit | |
Purified |
Flow Cytometry | |
70B1413.1 | |
Flow Cytometry, Flow (Intracellular) | |
activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6. | |
0.1 mL | |
Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
22926 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction