Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF6 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP175478B
Description
ATF6 Polyclonal specifically detects ATF6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
ATF6 | |
Polyclonal | |
Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence | |
P18850 | |
ATF6 | |
A genomic peptide made to an internal region of the human ATF6 protein (within residues 200-350). [Swiss-Prot P18850] | |
0.1 mL | |
Lipid and Metabolism, Signal Transduction, Unfolded Protein Response | |
22926 | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Biotin | |
PBS with 0.05% Sodium Azide | |
activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction