Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114886 Supplier Invitrogen™ Supplier No. PA5114886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATF6.

ATF6 is a transcription factor that acts during endoplasmic reticulum stress by activating unfolded protein response target genes. It binds DNA on the 5'-CCAC[GA]-3'half of the ER stress response element (ERSE) (5'-CCAAT-N(9)-CCAC[GA]-3') and of ERSE II (5'-ATTGG-N-CCACG-3'). Binding to ERSE requires binding of NF-Y to ERSE. ATF6 could also be involved in activation of transcription by the serum response factor. ATF6 exists as a homodimer and heterodimer with ATF6 beta. The dimer interacts with the nuclear transcription factor Y (NF-Y) trimer through direct binding to NF-Y subunit C (NF-YC). It Interacts also with the transcription factors GTF2I, YY1 and SRF. Under ER stress the cleaved N-terminal cytoplasmic domain translocates into the nucleus. The basic domain of ATF6 functions as a nuclear localization signal and the basic leucine-zipper domain is sufficient for association with the NF-Y trimer and binding to ERSE. During the unfolded protein response an approximately 50 kDa fragment containing the cytoplasmic transcription factor domain is released by proteolysis. The cleavage seems to be performed sequentially by site-1 and site-2 proteases. ATF6 is N-glycosylated, phosphorylated in vitro by MAPK14/P38MAPK and belongs to the bZIP family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ATF6
Gene Accession No. F6VAN0, G3V909, O35451, P18850, Q99941
Gene Alias 9130025P16Rik; 9630036G24; AA617266; AA789574; ACHM7; activating transcription factor 6; activating transcription factor 6 alpha; Activating transcription factor 6 beta; ATF 6; Atf6; ATF6 alpha; ATF6A; Atf6alpha; ATF6-alpha; Atf6b; ATF6beta; ATF6-beta; cAMP response element binding protein-related protein; cAMP response element-binding protein-related protein; cAMP responsive element binding protein-like 1; cAMP-dependent transcription factor ATF-6 alpha; cAMP-dependent transcription factor ATF-6 beta; cAMP-responsive element-binding protein-like 1; Crebl1; Creb-related protein; CREB-RP; Cyclic AMP-dependent transcription factor ATF-6 alpha; cyclic AMP-dependent transcription factor ATF-6 beta; DKFZp686P2194; ESTM49; FLJ21663; G13; Processed cyclic AMP-dependent transcription factor ATF-6 alpha; Processed cyclic AMP-dependent transcription factor ATF-6 beta; Protein G13
Gene Symbols ATF6, ATF6B
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATF6(Accession P18850), corresponding to amino acid residues T289-K339.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12915, 1388, 226641, 22926, 304962, 406169
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.