Learn More
Invitrogen™ ATF6 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Specifications
Specifications
| Antigen | ATF6 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ATF6 |
| Gene Accession No. | P18850, Q99941 |
| Gene Alias | 9130025P16Rik; 9630036G24; AA617266; AA789574; ACHM7; activating transcription factor 6; activating transcription factor 6 alpha; Activating transcription factor 6 beta; ATF 6; Atf6; ATF6 alpha; ATF6A; Atf6alpha; ATF6-alpha; Atf6b; ATF6beta; ATF6-beta; cAMP response element binding protein-related protein; cAMP response element-binding protein-related protein; cAMP responsive element binding protein-like 1; cAMP-dependent transcription factor ATF-6 alpha; cAMP-dependent transcription factor ATF-6 beta; cAMP-responsive element-binding protein-like 1; Crebl1; Creb-related protein; CREB-RP; Cyclic AMP-dependent transcription factor ATF-6 alpha; cyclic AMP-dependent transcription factor ATF-6 beta; DKFZp686P2194; ESTM49; FLJ21663; G13; Processed cyclic AMP-dependent transcription factor ATF-6 alpha; Processed cyclic AMP-dependent transcription factor ATF-6 beta; Protein G13 |
| Gene Symbols | ATF6, ATF6B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.