Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF6 Rabbit anti-Human, Clone: S05-4E7, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | ATF6 |
---|---|
Clone | S05-4E7 |
Dilution | Western Blot 1:100-1:2000, ELISA, Immunohistochemistry |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
ATF6 Monoclonal antibody specifically detects ATF6 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
ATF6 | |
Western Blot 1:100-1:2000, ELISA, Immunohistochemistry | |
Monoclonal | |
Purified | |
RUO | |
Human | |
activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
Recombinant protein of human ATF6 (Uniprot#: P18850) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S05-4E7 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
22926 | |
IgG | |
Affinity purified | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title