Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF6 Rabbit anti-Human, Mouse, Rat, Clone: 2358C, R&D Systems™

Catalog No. MAB71527100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB71527100 100 μg
MAB71527SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB71527100 Supplier R&D Systems Supplier No. MAB71527100
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ATF6 Monoclonal antibody specifically detects ATF6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen ATF6
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2358C
Conjugate Unconjugated
Dilution Western Blot 0.05 μg/mL, Immunohistochemistry 0.3-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. &asterisk;Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P18850
Gene Alias ACHM7, activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6 alpha, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha
Host Species Rabbit
Immunogen E. coli-derived recombinant human ATF6, Met1-Thr192, Accession No. P18850
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 22926.0
Test Specificity Detects human ATF6 in direct ELISAs. Detects human, mouse, and rat ATF6 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.