Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATF6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109425 Supplier Invitrogen™ Supplier No. PA5109425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAELMLLSEI ADPTRFFTDN LLSPEDWGLQ NSTLYSGLDE VAEEQTQLFR CPEQDVPFDG SSLDVGMDVS.

ATF6 beta(CREBL1) is a transcription factor in the unfolded protein response (UPR) pathway during ER stress. Either as a homodimer or as a heterodimer with ATF6-alpha, the protein binds to the ER stress response element, interacting with nuclear transcription factor Y to activate UPR target genes. ATF6 beta is normally found in the membrane of the endoplasmic reticulum; however, under ER stress, the N-terminal cytoplasmic domain is cleaved from the rest of the protein and translocates to the nucleus. Two transcript variants encoding different isoforms have been found for this gene. The protein encoded by this gene bears sequence similarity with the Creb/ATF subfamily of the bZip superfamily of transcription factors. It localizes to both the cytoplasm and the nucleus. The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF6B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Atf6b
Gene Accession No. O35451
Gene Alias AA617266; Activating transcription factor 6 beta; ATF6B; ATF6beta; ATF6-beta; cAMP response element binding protein-related protein; cAMP response element-binding protein-related protein; cAMP responsive element binding protein-like 1; cAMP-dependent transcription factor ATF-6 beta; cAMP-responsive element-binding protein-like 1; CREBL1; Creb-related protein; Creb-rp; cyclic AMP-dependent transcription factor ATF-6 beta; FLJ10066; G13; Processed cyclic AMP-dependent transcription factor ATF-6 beta; Protein G13
Gene Symbols Atf6b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human ATF6B (Q99941).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12915, 406169
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.