Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF7 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130071AF750
Description
ATF7 Polyclonal antibody specifically detects ATF7 in Human samples. It is validated for Western BlotSpecifications
ATF7 | |
Polyclonal | |
Western Blot | |
activating transcription factor 7ATFATranscription factor ATF-A, cAMP-dependent transcription factor ATF-7, cyclic AMP-dependent transcription factor ATF-7, MGC57182 | |
maps to a region between residue 444 and 494 of human activating transcription factor 7 using the numbering given in entry CAA40483.1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
11016 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction