Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF7 Antibody [DyLight 550], Novus Biologicals Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP274750H
Description
ATF7 Polyclonal antibody specifically detects ATF7 in Human samples. It is validated for ELISA,Western BlotSpecifications
ATF7 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
11016 | |
Store at 4°C in the dark. | |
Purified |
ELISA, Western Blot | |
DyLight 550 | |
activating transcription factor 7ATFATranscription factor ATF-A, cAMP-dependent transcription factor ATF-7, cyclic AMP-dependent transcription factor ATF-7, MGC57182 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human ATF7 (NP_001193611.1).,, Sequence:, MGDDRPFVCNAPGCGQRFTNEDHLAVHKHKHEMTLKFGPARTDSVIIADQTPTPTRFLKNCEEVGLFNELASSFEHEFKKAADEDEKKARSRTVAKKLVVFRPRLFLLCFGIIFLIG | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction