Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATF7IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568833 Supplier Invitrogen™ Supplier No. PA568833
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Pig: 92%; Rat: 85%.

ATF7IP is a multifunctional nuclear protein that associates with heterochromatin. It can act as a transcriptional coactivator or corepressor depending upon its binding partners.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF7IP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Atf7ip
Gene Accession No. Q7TT18
Gene Alias 2610204M12Rik; activating transcription factor 7 interacting protein; activating transcription factor 7-interacting protein 1; AM; ATF7-interacting protein; ATF7IP; ATFa-associated modulator; ATF-interacting protein; ATF-IP; FLJ10688; hAM; mAM; MBD1-containing chromatin-associated factor 1; MCAF; MCAF1; p621
Gene Symbols Atf7ip
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse ATF7IP.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54343
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.