Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG1/ULK1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985524
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5524 Supplier Bioss Supplier No. BS3602R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ULK1, also known as ATG1, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK1 results in a severe defect in the autophagy pathway. ULK1 is highly conserved among eukaryotes, and are the Unc-51-like kinases, ULK1 and ULK2 in mammals. ULK1 is ubiquitously expressed and involved in autophagy in response to starvation. It is the target of the TOR kinase signaling pathway that regulates autophagy through the control of phosphorylation status of ATG13. ULK1 also plays a role early in neuronal differentiation.

Specifications

Antigen ATG1/ULK1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ULK1
Gene Accession No. O75385
Gene Alias APG1; ATG 1; ATG1; ATG1 autophagy related 1 homolog; ATG1A; AU041434; Autophagy-related protein 1 homolog; C. elegans; EC:2.7.11.1; FLJ38455; hATG1; KIAA0722; mKIAA0722; Serine/threonine-protein kinase ULK1; Serine/threonine-protein kinase Unc51.1; ULK 1; ULK1; UNC 51; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like kinase 1; Unc51.1; Unc-51-like kinase 1
Gene Symbols ULK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATG1/ULK1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8408
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.