Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG10 Antibody (SR1389), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32173050UL
Description
ATG10 Monoclonal antibody specifically detects ATG10 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ATG10 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR1389 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10 | |
A synthesized peptide derived from human ATG10 (Uniprot #: Q9H0Y0) | |
50 μg | |
Autophagy | |
83734 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction