Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523224
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523224 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523224 Supplier Invitrogen™ Supplier No. PA523224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG10 Polyclonal Antibody for Western Blot, IHC (P)

ATG10 is involved in autophagy which is a process for the bulk degradation of cytosolic compartments by lysosomes/vacuoles. The formation of autophagosomes involves a dynamic rearrangement of the membrane for which two ubiquitin-like modifications (the conjugation of APG12p and the modification of a soluble form of MAP-LC3 to a membrane-bound form) are essential. APG10 is an E2-like enzyme involved in two ubiquitin-like modifications essential for autophagosome formation, ATG12 -ATG5 conjugation and modification of a soluble form of MAP-LC3 (MAP1LC3A), a homolog of yeast Apg8, to a membrane-bound form. The protein ATG10 is ubiquitously expressed and the human APG10L gene is localized on chromosome 5q14.2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene ATG10
Gene Accession No. Q8R1P4, Q9H0Y0
Gene Alias 5330424L23Rik; 5430428K15Rik; Agp10; AI852123; APG10; APG10L; APG10-like; Apg10p; ATG10; ATG10 autophagy related 10 homolog; Atg10l; autophagy related 10; autophagy related 10 homolog; autophagy-related 10; autophagy-related protein 10; DKFZp586I0418; FLJ13954; mAPG10; pp12616; Ubiquitin-like-conjugating enzyme ATG10
Gene Symbols ATG10
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human APG10L.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66795, 688555, 83734
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.