Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521000
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521000 Supplier Thermo Scientific Supplier No. PA521000
Add to Cart
Add to Cart

ATG101 Polyclonal Antibody

ATG101 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG101
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ATG101
Gene Accession No. Q6AY69, Q9BSB4, Q9D8Z6
Gene Alias 9430023L20Rik; AA407441; ATG101; Atg13-interacting protein; AU022967; autophagy related 101; autophagy-related protein 101; C12orf44; PP894; RGD1359310
Gene Symbols ATG101
Host Species Rabbit
Immunogen A 16 amino acid peptide near the center of human ATG101.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300240, 60673, 68118
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.