Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520464
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520464 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520464 Supplier Invitrogen™ Supplier No. PA520464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse heart tissue lysate. PA5-20464 can be used with blocking peptide PEP-0581.

ATG12, a member of the autophagy protein family, forms a conjugate with ATG5; this conjugate has a ubiquitin-protein ligase (E3)-like activity for protein lipidation in autophagy. This conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. ATG12 has also been shown to interact with ATG10 in human embryonic kidney cells in the presence of ATG7. At least two isoforms of ATG12 are known to exist. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG12
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ATG12
Gene Accession No. O94817, Q2TBJ5, Q9CQY1
Gene Alias 4931423H11Rik; A330058M13Rik; APG 12; APG12; Apg12 (autophagy, yeast) homolog; APG12L; APG12-like; ATG12; ATG12 autophagy related 12 homolog; Atg12l; Atg12p; Atg12-PB; Autophagy protein 12-like; autophagy related 12; autophagy related protein 12; autophagy-related 12; autophagy-related protein 12; Autophagy-specific gene 12; CG10861; CG10861-PB; Dmel\CG10861; Dmel_CG10861; FBR93; HAPG12; IP05205p; IP05405p; OTTHUMP00000159129; ubiquitin-like protein ATG12; YBR1506; YBR217W
Gene Symbols ATG12
Host Species Rabbit
Immunogen A 15 amino acid peptide from near the center of human ATG12.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361321, 67526, 9140
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.