Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atg12 Polyclonal Antibody, Huabio

Catalog No. 502544714
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4714 Supplier HUABIO RESEARCH INC Supplier No. ER180282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Atg12 Polyclonal antibody specifically detects Atg12 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Atg12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ATG12
Gene Accession No. O94817
Gene Alias 4931423H11Rik; A330058M13Rik; APG 12; APG12; Apg12 (autophagy, yeast) homolog; APG12L; APG12-like; ATG12; ATG12 autophagy related 12 homolog; Atg12l; Atg12p; Atg12-PB; Autophagy protein 12-like; autophagy related 12; autophagy related protein 12; autophagy-related 12; autophagy-related protein 12; Autophagy-specific gene 12; CG10861; CG10861-PB; Dmel\CG10861; Dmel_CG10861; FBR93; HAPG12; IP05205p; IP05405p; OTTHUMP00000159129; ubiquitin-like protein ATG12; YBR1506; YBR217W
Host Species Rabbit
Immunogen Recombinant protein within human Atg12 aa 1-120.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.