Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243577
Description
Specifically detects ATG14 in Human samples, and it is validated for Western Blotting
ATG14 Polyclonal specifically detects ATG14 in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
ATG14 | |
Polyclonal | |
Western Blot 1:500-1:3000, Knockout Validated | |
ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831 | |
Rabbit | |
55 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Immunoassay | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
ATG14 | |
Recombinant protein encompassing a sequence within the center region of human ATG14. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
22863 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction