Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985829
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5829 Supplier Bioss Supplier No. BS4007R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Autophagy related protein 16 L is encoded by the ATG16L gene and is located on chromosome 2 in humans. It is also known as WD30 or IBD10 (Inflammatory Bowel Disease 10). ATG 12 and ATG5 conjugate with each other and undergoes homo oligomerisation with ATG16L to form an 800 kDa complex called the Atg16L complex. This complex facilitates coupling or lipidation of LC3 to phosphatidylethanolamine (PE) and this ubiquitin-like conjugation system is a widely used marker for autophagy. However, the ATG 16L complex along with other ATGs like ATG 18 and ATG 21 prevent the premature cleavage of ATG8-PE by the cysteine protease, Atg4 until the autophagosome is deemed complete.

Specifications

Antigen ATG16L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATG16L1
Gene Accession No. Q676U5
Gene Alias 1500009K01Rik; A16L1; APG 16-L1; Apg16l; APG16L beta; APG16L isoforms 1; APG16-like 1; ATG16 autophagy related 16-like 1; ATG16A; ATG16L; ATG16L1; Atg16L1 gamma; autophagy related 16 like 1; autophagy related 16-like 1; autophagy related 16-like 1 (S. cerevisiae); autophagy-related 16-like 1; autophagy-related 16-like 1 gamma; Autophagy-related protein 16-1; hCG_1817841; IBD10; UNQ9393/PRO34307; WD repeat domain 30; Wdr30
Gene Symbols ATG16L1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATG16A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55054
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.