Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 9 publications

Supplier:  Novus Biologicals NB11060928SS

 View more versions of this product

Catalog No. NB11060928T


Only null left
Add to Cart

Description

Description

ATG16L1 Polyclonal specifically detects ATG16L1 in Human, Mouse, Rat, Alligator, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy.
Specifications

Specifications

ATG16L1
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.1% Sodium Azide
APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30
Rabbit
68 kDa
0.025 mL
Autophagy, Cancer, Cell Biology, Macroautophagy, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
55054
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
0.8 mg/mL
Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25060858), Electron Microscopy 1:10-1:500. Use reported in scientific literature (PMID 22531915)
Q676U5
ATG16L1
A synthetic peptide within residues 1-100 of human ATG16L1 protein. [Swiss-Prot# Q676U5]
Affinity Purified
RUO
Primary
Human, Mouse, Rat, Alligator, Bovine, Canine, Primate
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only