Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG16L1 Monoclonal Antibody (OTI2A10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809571

Description
Autophagy related protein 16 L is encoded by the ATG16L gene and is located on chromosome 2 in humans. It is also known as WD30 or IBD10 (Inflammatory Bowel Disease 10). ATG 12 and ATG5 conjugate with each other and undergoes homo oligomerisation with ATG16L to form an 800 kDa complex called the Atg16L complex. This complex facilitates coupling or lipidation of LC3 to phosphatidylethanolamine (PE) and this ubiquitin-like conjugation system is a widely used marker for autophagy. However, the ATG 16L complex along with other ATGs like ATG 18 and ATG 21 prevent the premature cleavage of ATG8-PE by the cysteine protease, Atg4 until the autophagosome is deemed complete.Specifications
ATG16L1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q676U5 | |
ATG16L1 | |
Human recombit protein fragment corresponding to amino acids 1-277 of human ATG16L produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI2A10 | |
Unconjugated | |
ATG16L1 | |
APG16L, ATG16A, ATG16L, IBD10, WDR30 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55054 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction