Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG16L1 Mouse anti-Human, Clone:8F8C2, Abnova™
Mouse monoclonal antibody raised against recombinant human ATG16L1.
Supplier: Abnova Corporation MAB17120
Description
Autophagy is the major intracellular degradation system delivering cytoplasmic components to lysosomes, and it accounts for degradation of most long-lived proteins and some organelles. Cytoplasmic constituents, including organelles, are sequestered into double-membraned autophagosomes, which subsequently fuse with lysosomes. ATG16L1 is a component of a large protein complex essential for autophagy (Mizushima et al., 2003 [PubMed 12665549]).[supplied by OMIM]Specifications
ATG16L1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
ATG16L1 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
8F8C2 | |
ATG16 autophagy related 16-like 1 (S. cerevisiae) | |
In PBS (0.05% sodium azide) | |
APG16L, ATG16L, FLJ00045, FLJ10035, FLJ10828, FLJ22677, IBD10, WDR30 | |
Recombinant protein corresponding to amino acid 11-257 of human ATG16L1 from E. coli. | |
55054 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction