Learn More
Invitrogen™ ATG16L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595969
Description
Immunogen sequence: IKHQEELTEL HKKRGELAQL VIDLNNQMQR KDREMQMNEA KIAECLQTIS DLETECLDLR TKLCDLERAN QTLKDEYDAL QITFTALEGK LRKTTEENQE LVTRWMAEKA QEANRLNAEN EKDSRRRQAR LQKELAEAAK EPLPVEQDDD IEVIVDETSD HTEETSPVRA ISRAATKRLS QPAGGLLDSI TNIFGRRSVS SFPVPQDNVD T; Positive Samples: U-937, 293T, HepG2, SH-SY5Y, SW620; Cellular Location: Cytoplasm, Peripheral membrane protein, Preautophagosomal structure membrane.
Autophagy related protein 16 L is encoded by the ATG16L gene and is located on chromosome 2 in humans. It is also known as WD30 or IBD10 (Inflammatory Bowel Disease 10). ATG 12 and ATG5 conjugate with each other and undergoes homo oligomerisation with ATG16L to form an 800 kDa complex called the Atg16L complex. This complex facilitates coupling or lipidation of LC3 to phosphatidylethanolamine (PE) and this ubiquitin-like conjugation system is a widely used marker for autophagy. However, the ATG 16L complex along with other ATGs like ATG 18 and ATG 21 prevent the premature cleavage of ATG8-PE by the cysteine protease, Atg4 until the autophagosome is deemed complete.
Specifications
ATG16L1 | |
Polyclonal | |
Unconjugated | |
ATG16L1 | |
1500009K01Rik; A16L1; APG 16-L1; Apg16l; APG16L beta; APG16L isoforms 1; APG16-like 1; atg16; ATG16 autophagy related 16-like 1; ATG16A; Atg16l; Atg16l1; Atg16L1 gamma; autophagy related 16 like 1; autophagy related 16-like 1; autophagy related 16-like 1 (S. cerevisiae); autophagy-related 16-like 1; autophagy-related 16-like 1 gamma; autophagy-related protein 16-1; hCG_1817841; IBD10; UNQ9393/PRO34307; WD repeat domain 30; Wdr30 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
363278, 55054, 77040 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.06 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q676U5, Q8C0J2 | |
ATG16L1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 90-300 of human ATG16L1 (NP_1104305). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.