Learn More
Invitrogen™ ATG16L1 Polyclonal Antibody

Description
Suggested positive control: NIH-3T3 whole cell lysates.
Specifications
Specifications
| Antigen | ATG16L1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | ATG16L1 |
| Gene Accession No. | Q676U5, Q8C0J2 |
| Gene Alias | 1500009K01Rik; A16L1; APG 16-L1; Apg16l; APG16L beta; APG16L isoforms 1; APG16-like 1; atg16; ATG16 autophagy related 16-like 1; ATG16A; Atg16l; Atg16l1; Atg16L1 gamma; autophagy related 16 like 1; autophagy related 16-like 1; autophagy related 16-like 1 (S. cerevisiae); autophagy-related 16-like 1; autophagy-related 16-like 1 gamma; autophagy-related protein 16-1; hCG_1817841; IBD10; UNQ9393/PRO34307; WD repeat domain 30; Wdr30 |
| Gene Symbols | ATG16L1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.