Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG16L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535207
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535207 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535207 Supplier Invitrogen™ Supplier No. PA535207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG16L1 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

Autophagy related protein 16 L is encoded by the ATG16L gene and is located on chromosome 2 in humans. It is also known as WD30 or IBD10 (Inflammatory Bowel Disease 10). ATG 12 and ATG5 conjugate with each other and undergoes homo oligomerisation with ATG16L to form an 800 kDa complex called the Atg16L complex. This complex facilitates coupling or lipidation of LC3 to phosphatidylethanolamine (PE) and this ubiquitin-like conjugation system is a widely used marker for autophagy. However, the ATG 16L complex along with other ATGs like ATG 18 and ATG 21 prevent the premature cleavage of ATG8-PE by the cysteine protease, Atg4 until the autophagosome is deemed complete.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG16L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ATG16L1
Gene Accession No. Q676U5, Q8C0J2
Gene Alias 1500009K01Rik; A16L1; APG 16-L1; Apg16l; APG16L beta; APG16L isoforms 1; APG16-like 1; atg16; ATG16 autophagy related 16-like 1; ATG16A; Atg16l; Atg16l1; Atg16L1 gamma; autophagy related 16 like 1; autophagy related 16-like 1; autophagy related 16-like 1 (S. cerevisiae); autophagy-related 16-like 1; autophagy-related 16-like 1 gamma; autophagy-related protein 16-1; hCG_1817841; IBD10; UNQ9393/PRO34307; WD repeat domain 30; Wdr30
Gene Symbols ATG16L1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 161-190 amino acids from human ATG16L.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55054, 77040
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.