Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATG16L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550183
Description
The antibody detects endogenous levels of total ATG16L2 protein.
May play a role in autophagy. [UniProt].
Specifications
ATG16L2 | |
Polyclonal | |
Unconjugated | |
ATG16L2 | |
2410118P20Rik; APG16-like 2; ATG16 autophagy related 16-like 2; ATG16B; Atg16l2; Atg16L2 alpha; Atg16L2 beta; autophagy related 16 like 2; autophagy related 16-like 2; autophagy related 16-like 2 (S. cerevisiae); autophagy-related 16-like 2 alpha; autophagy-related 16-like 2 beta; autophagy-related protein 16-2; mFLJ00012 protein; RGD1311400; WD repeat-containing protein 80; WDR80 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
73683, 89849 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6KAU8, Q8NAA4 | |
ATG16L2 | |
Synthesized peptide derived from internal of human ATG16L2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction