Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG2A Antibody (4E6D4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261690
Description
ATG2A Monoclonal specifically detects ATG2A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ATG2A | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
ATG2 autophagy related 2 homolog A (S. cerevisiae), autophagy-related protein 2 homolog A, KIAA0404MGC117153 | |
Mouse | |
212.8 kDa | |
0.1 mL | |
23130 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
4E6D4 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ATG2A | |
Purified recombinant fragment of human ATG2A (AA: 325-429) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction