Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATG2A Monoclonal Antibody (4H8G3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531638
Description
MA5-31638 has been tested in indirect ELISA.
Involved in autophagosome assembly, regulating the size of nascent autophagosomes (PubMed:28561066). Also regulates lipid droplets morphology and distribution within the cell (PubMed:22219374, PubMed:28561066). [UniProt].
Specifications
ATG2A | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q2TAZ0 | |
ATG2A | |
Purified recombinant fragment of human ATG2A (AA: 325-429) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
4H8G3 | |
Unconjugated | |
ATG2A | |
1810013C15Rik; A830054M12; ATG2 autophagy related 2 homolog A; Atg2a; autophagy related 2A; autophagy-related 2-like protein A; autophagy-related protein 2 homolog A; BC023754; Kiaa0404; mKIAA0404 | |
Mouse | |
Protein G | |
RUO | |
23130 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction