Learn More
Invitrogen™ ATG2A Polyclonal Antibody

Description
Immunogen sequence: EDLWLIEQDL NQQLQAGAVA EPLSPDPLTN PLLNLDNTDL FFSMAGLTSS VASALSELSL SDVDLASSVR SDMASRRLSA QAHPAGKMAP NPLLDTMRPD SLLKM Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
Specifications
Specifications
| Antigen | ATG2A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ATG2A |
| Gene Accession No. | Q2TAZ0 |
| Gene Alias | 1810013C15Rik; A830054M12; ATG2 autophagy related 2 homolog A; Atg2a; autophagy related 2A; autophagy-related 2-like protein A; autophagy-related protein 2 homolog A; BC023754; Kiaa0404; mKIAA0404 |
| Gene Symbols | ATG2A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.