Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578435 Supplier Invitrogen™ Supplier No. PA578435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: U87-MG, SK-N-SH Predicted Reactivity: Mouse (88%), Rat (88%), Pig (91%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Involved in autophagosome assembly, regulating the size of nascent autophagosomes (PubMed:28561066). Also regulates lipid droplets morphology and distribution within the cell (PubMed:22219374, PubMed:28561066). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG2A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ATG2A
Gene Accession No. Q2TAZ0
Gene Alias 1810013C15Rik; A830054M12; ATG2 autophagy related 2 homolog A; Atg2a; autophagy related 2A; autophagy-related 2-like protein A; autophagy-related protein 2 homolog A; BC023754; Kiaa0404; mKIAA0404
Gene Symbols ATG2A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ATG2A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23130
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.