Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597122 Supplier Invitrogen™ Supplier No. PA597122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLHGIYEDGG KPPVPCLRVS KALDPKSTGR KYFLPQVVVT VNPQSSSTQW EVAPEKGEEL ELSVESPCEL REPEPSPFSS KRTMYETEEM VIPGDPEEMR T; Positive Samples: U-87MG, HeLa; Cellular Location: Lipid droplet, Peripheral membrane protein, Preautophagosomal structure membrane.

Involved in autophagosome assembly, regulating the size of nascent autophagosomes (PubMed:28561066). Also regulates lipid droplets morphology and distribution within the cell (PubMed:22219374, PubMed:28561066). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG2A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATG2A
Gene Accession No. Q2TAZ0
Gene Alias 1810013C15Rik; A830054M12; ATG2 autophagy related 2 homolog A; Atg2a; autophagy related 2A; autophagy-related 2-like protein A; autophagy-related protein 2 homolog A; BC023754; Kiaa0404; mKIAA0404
Gene Symbols ATG2A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 700-800 of human ATG2A (NP_0559192).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23130
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.